4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO) 98+% free radical
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23456-03
B23456-06
CAAAB23456-03EA
78.21
CAD
CAAAB23456-03
CAAAB23456-06
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO) 98+% free radical
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-Acetamido-TEMPO, free radical)
Purity: 98+%
CAS No.: 14691-89-5
Molecular Formula: C11H21N2O2
Formula Weight: 213.30
Storage Temperature: 2°C to 8°C
Melting Point: 143-146°
Beilstein Reference: 3546225
MDL No.: MFCD00043593
Formula:
C₁₁H₂₁N₂O₂ Melting Pt: 143…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043593 CAS Number: 14691-89-5 EINECS: 423-840-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...