4-(Methylthio)phenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Methylthio)benzeneboronic acid, 4-Thioanisoleboronic acid
B23454-03
B23454-06
B23454-14
CAAAB23454-03EA
44.71
CAD
CAAAB23454-03
CAAAB23454-06
CAAAB23454-14
4-(Methylthio)phenylboronic acid 97%
4-(Methylthio)phenylboronic acid
Purity: 97%
CAS No.: 98546-51-1
Molecular Formula: C7H9BO2S
Formula Weight: 168.02
Melting Point: ca 208-212°
Beilstein Reference: 3247219
Formula:
C₇H₉BO₂S Melting Pt: ca 208…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00093410 CAS Number: 98546-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...