Piperidine-N-carbaldehyde 99%

Supplier: Thermo Scientific Chemicals
Danger

B23452-0B B23452-18 B23452-30
CAAAB23452-0BEA 535.62 CAD
CAAAB23452-0B CAAAB23452-18 CAAAB23452-30
Piperidine-N-carbaldehyde 99%
Piperidine-N-carbaldehyde

Purity: 99% CAS No.: 2591-86-8 Molecular Formula: C6H11NO Formula Weight: 113.16 Flash Point: 102°(215°F) Refractive Index: 1.4840 Melting Point: -31° Boiling Point: 222° Beilstein Reference: 107697 MDL No.: MFCD00006483

Formula: C₆H₁₁NO
Boiling Pt: 222 °C
Melting Pt: –31 °C
Density: 1.019
Flash Pt: 102 °C(215 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006483
CAS Number: 2591-86-8
EINECS: 219-986-0
UN: 2810
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR