4-Cumylphenol 97%
Supplier: Thermo Scientific Chemicals
B23410-22
B23410-0B
B23410-30
CAAAAB23410-22EA
76.79
CAD
CAAAAB23410-22
CAAAB23410-0B
CAAAB23410-30
4-Cumylphenol 97%
4-Cumylphenol
Purity: 97%
CAS No.: 599-64-4
Molecular Formula: C15H16O
Formula Weight: 212.29
Flash Point: 160°(320°F)
Melting Point: 70-73°
Boiling Point: 189-191°/10mm
Beilstein Reference: 1870517
MDL No.: MFCD00002365
Formula:
C₁₅H₁₆O Boiling Pt: 189…191 °C/10 mm Melting Pt: 70…73 °C Density: 1.115 Flash Pt: 160 °C(320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002365 CAS Number: 599-64-4 EINECS: 209-968-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...