5-Methyl-2-thiopheneboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23138-03
B23138-06
CAAAB23138-03EA
79
CAD
CAAAB23138-03
CAAAB23138-06
CAAAB23138-14
5-Methyl-2-thiopheneboronic acid 98%
5-Methyl-2-thiopheneboronic acid
Purity: 98%
CAS No.: 162607-20-7
Molecular Formula: C5H7BO2S
Formula Weight: 142.01
Storage Temperature: 2°C to 8°C
Melting Point: ca 160° dec.
Beilstein Reference: 7019569
MDL No.: MFCD01318166
Formula:
C₅H₇BO₂S Melting Pt: ∼ 160 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01318166 CAS Number: 162607-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...