Trimellitic anhydride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2,4-Benzenetricarboxylic anhydride, 1,2,4-Benzenetricarboxylic 1,2-anhydride

B23003-30 B23003-A1
CAAAB23003-30EA 50.56 CAD
CAAAB23003-30 CAAAB23003-A1
Trimellitic anhydride 97%
Trimellitic anhydride

Purity: 97% CAS No.: 552-30-7 Molecular Formula: C9H4O5 Formula Weight: 192.13 Flash Point: 227°(440°F) Melting Point: 165-169° Boiling Point: 240-245°/14mm Beilstein Reference: 9394 Merck Reference: 14,9703 MDL No.: MFCD00005925

Formula: C₉H₄O₅
MW: 192.13 g/mol
Boiling Pt: 240…245 °C/14 mm
Melting Pt: 165…169 °C
Density: 1.540
Flash Pt: 227 °C(440 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005925
CAS Number: 552-30-7
EINECS: 209-008-0
Merck Index: 14,9703

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR