1-(2-Chloroethyl)piperidinium chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(2-Chloroethyl)-piperidine hydrochloride, 1-(2-Chloroethyl)piperidine hydrochloride

B22985-18 B22985-30
CAAAAB22985-36EA 330.33 CAD
CAAAAB22985-36 CAAAB22985-18 CAAAB22985-30
1-(2-Chloroethyl)piperidinium chloride 98%
1-(2-Chloroethyl)piperidinium chloride

Purity: 98% CAS No.: 2008-75-5 Molecular Formula: C7H14ClN.HCl Formula Weight: 184.11 Melting Point: 229-235° Beilstein Reference: 3910268 UN No.: 2928 MDL No.: MFCD00012837

Formula: C₇H₁₅Cl₂N
MW: 184.11 g/mol
Melting Pt: 229…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00012837
CAS Number: 2008-75-5
EINECS: 217-920-5
UN: 2928
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR