3,3-Diphenylpropylamine 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,3-Diphenylpropan-1-amine

B22953-14 B22953-22
CAAAB22953-14EA 122.61 CAD
CAAAB22953-14 CAAAB22953-22
3,3-Diphenylpropylamine 97%
3,3-Diphenylpropylamine

Purity: 97% CAS No.: 5586-73-2 Molecular Formula: (C6H5)2CHCH2CH2NH2 Formula Weight: 211.31 Flash Point: 110°(230°F) Refractive Index: 1.5830 Melting Point: 24-29° Boiling Point: 166°/2mm Beilstein Reference: 1958595 MDL No.: MFCD00008202

Formula: C₁₅H₁₇N
MW: 211.31 g/mol
Boiling Pt: 166 °C/2 mm
Melting Pt: 24…29 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008202
CAS Number: 5586-73-2
EINECS: 226-984-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR