Diethyl benzylmalonate 97%

Supplier: Thermo Scientific Chemicals

B22890-14 B22890-22
CAAAB22890-14EA 55.93 CAD
CAAAB22890-14 CAAAB22890-22
Diethyl benzylmalonate 97%
Diethyl benzylmalonate

Purity: 97% CAS No.: 607-81-8 Molecular Formula: C14H18O4 Formula Weight: 250.29 Flash Point: 110°(230°F) Refractive Index: 1.4860 Boiling Point: 162-163°/10mm Beilstein Reference: 615793 MDL No.: MFCD00009166

Formula: C₁₄H₁₈O₄
Boiling Pt: 162…163 °C/10 mm
Density: 1.069
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009166
CAS Number: 607-81-8
EINECS: 210-142-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR