Disodium benzene-1,3-disulfonate 94%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3-Benzenedisulfonic acid disodium salt, 1,3-Benzenedisulphonic acid disodium salt, Benzene-1,3-disulfonic acid disodium salt, Benzene-1,3-disulphonic acid disodium salt

B22811-14 B22811-22 B22811-36
CAAAB22811-14EA 56.56 CAD
CAAAB22811-14 CAAAB22811-22 CAAAB22811-36
Disodium benzene-1,3-disulfonate 94%
Disodium benzene-1,3-disulphonate

Purity: 94% CAS No.: 831-59-4 Molecular Formula: C6H4Na2O6S2 Formula Weight: 282.20 MDL No.: MFCD00007492

Formula: C₆H₄Na₂O₆S₂
Density: 1,124
Storage Temperature: Ambient
MDL Number: MFCD00007492
CAS Number: 831-59-4
EINECS: 212-606-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR