N-(2-Hydroxyethyl)morpholine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(2-Hydroxyethyl)morpholine, 2-Morpholinoethanol

B22759-30 B22759-22 B22759-36
CAAAAB22759-30EA 63.2 CAD
CAAAAB22759-30 CAAAB22759-22 CAAAB22759-36
N-(2-Hydroxyethyl)morpholine 99%
N-(2-Hydroxyethyl)morpholine

Purity: 99% CAS No.: 622-40-2 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Flash Point: 99°(210°F) Refractive Index: 1.4775 Melting Point: 1-2° Boiling Point: 226-228° Beilstein Reference: 104375 MDL No.: MFCD00006180

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 226…228 °C
Melting Pt: 1…2 °C
Density: 1.080
Flash Pt: 99 °C(210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006180
CAS Number: 622-40-2
EINECS: 210-734-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR