2,2,6,6-Tetramethylpiperidin-4-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2,6,6-Tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethylpiperidine
B22664-06
B22664-14
B22664-22
CAAAB22664-06EA
54.19
CAD
CAAAB22664-06
CAAAB22664-14
CAAAB22664-22
2,2,6,6-Tetramethylpiperidin-4-ol 98%
2,2,6,6-Tetramethylpiperidin-4-ol
Purity: 98%
CAS No.: 2403-88-5
Molecular Formula: C9H19NO
Formula Weight: 157.26
Flash Point: 124°(255°F)
pH: 11.2
Melting Point: 129-133°
Boiling Point: 212-215°
Beilstein Reference: 105039
UN No.: 3259
MDL No.: MFCD00005983
Formula:
C₉H₁₉NO MW: 157.26 g/mol Boiling Pt: 212…215 °C Melting Pt: 129…133 °C Density: 1.085 Flash Pt: 124 °C(255 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005983 CAS Number: 2403-88-5 EINECS: 219-291-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...