2-Chloroethyldiisopropylammonium chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

B22662-22 B22662-36
CAAAB22662-0EEA 1415.3 CAD
CAAAB22662-0E CAAAB22662-22 CAAAB22662-36
2-Chloroethyldiisopropylammonium chloride 98%
2-Chloroethyldiisopropylammonium chloride

Purity: 98% CAS No.: 4261-68-1 Molecular Formula: [(CH3)2CH]2NCH2CH2Cl.HCl Formula Weight: 200.15 Melting Point: 131-134° Beilstein Reference: 3551603 UN No.: 2811 MDL No.: MFCD00012496

Formula: C₈H₁₉Cl₂N
Melting Pt: 131…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00012496
CAS Number: 4261-68-1
EINECS: 224-238-1
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR