Dibenzyl diisopropylphosphoramidite 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dibenzyl N,N-diisopropylphosphoramidite
B22629-06
B22629-14
B22629-22
CAAAB22629-06EA
148.2
CAD
CAAAB22629-06
CAAAB22629-14
CAAAB22629-22
Dibenzyl diisopropylphosphoramidite 94%
Dibenzyl diisopropylphosphoramidite
Purity: 95%
CAS No.: 108549-23-1
Molecular Formula: C20H28NO2P
Formula Weight: 345.43
Storage Temperature: 2°C to 8°C
Flash Point: 69°(156°F)
Physical Form: Liquid
Density: 1.030
Sensitivity: Moisture Sensitive
Boiling Point: 130°/0.55mm
MDL No.: MFCD00191988
Formula:
C₂₀H₂₈NO₂P Boiling Pt: 130 °C/0.55 mm Density: 1.030 Flash Pt: 69 °C(156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00191988 CAS Number: 108549-23-1 EINECS: 629-628-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...