Ethylphosphonic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22559-06
B22559-14
CAAAB22559-06EA
276.5
CAD
CAAAB22559-06
CAAAB22559-14
Ethylphosphonic acid 98+%
Ethylphosphonic acid
Purity: 98+%
CAS No.: 6779-09-5
Molecular Formula: C2H7O3P
Formula Weight: 110.06
Melting Point: 58-62°
Beilstein Reference: 1743032
UN No.: 3261
MDL No.: MFCD00002141
Formula:
C₂H₇O₃P Melting Pt: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002141 CAS Number: 6779-09-5 EINECS: 229-839-2 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...