Methyl-2,5-dichlorobenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2,5-dichlorobenzoate

B22489-09 B22489-18
CAAAB22489-09EA 85.64 CAD
CAAAB22489-09 CAAAB22489-18
Methyl-2,5-dichlorobenzoate 99%
Methyl 2,5-dichlorobenzoate

Purity: 99% CAS No.: 2905-69-3 Molecular Formula: C8H6Cl2O2 Formula Weight: 205.04 Flash Point: 110°(230°F) Melting Point: 37-40° Beilstein Reference: 2251582 MDL No.: MFCD00000606

Formula: C₈H₆Cl₂O₂
MW: 205.04 g/mol
Melting Pt: 37…40 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000606
CAS Number: 2905-69-3
EINECS: 220-815-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR