1-Methyl-1-phenylhydrazine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22440-06
B22440-14
B22440-22
CAAAB22440-06EA
181.7
CAD
CAAAB22440-06
CAAAB22440-14
CAAAB22440-22
1-Methyl-1-phenylhydrazine 97%
1-Methyl-1-phenylhydrazine
Purity: 97%
CAS No.: 618-40-6
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 92°(198°F)
Refractive Index: 1.5830
Boiling Point: 54-55°/0.3mm
MDL No.: MFCD00007627
Formula:
C₇H₁₀N₂ Boiling Pt: 54…55 °C/0.3 mm Density: 1.038 Flash Pt: 92 °C(198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007627 CAS Number: 618-40-6 EINECS: 210-547-9 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...