Acetonyltriphenylphosphonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

B22434-06 B22434-14
CAAAB22434-06EA 96.22 CAD
CAAAB22434-06 CAAAB22434-14
Acetonyltriphenylphosphonium bromide 98+%
Acetonyltriphenylphosphonium bromide

Fieser: 21,1


Purity: 98+% CAS No.: 2236-01-3 Molecular Formula: C21H20BrOP Formula Weight: 399.27 Melting Point: 226-227° Beilstein Reference: 3802148

Formula: C₂₁H₂₀BrOP
Melting Pt: 226…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00051863
CAS Number: 2236-01-3
EINECS: 218-796-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR