1-Methyl-2-piperidinemethanol 98%

Supplier: Thermo Scientific Chemicals
Danger

B22187-06 B22187-14 B22187-22
CAAAB22187-06EA 42.03 CAD
CAAAB22187-06 CAAAB22187-14 CAAAB22187-22
1-Methyl-2-piperidinemethanol 98%
1-Methyl-2-piperidinemethanol

Purity: 98% CAS No.: 20845-34-5 Molecular Formula: C7H15NO Formula Weight: 129.20 Flash Point: 81°(177°F) Refractive Index: 1.4830 Boiling Point: 79-80°/7mm Beilstein Reference: 103612 MDL No.: MFCD00006494

Formula: C₇H₁₅NO
Boiling Pt: 79…80 °C/7 mm
Density: 0.984
Flash Pt: 81 °C(177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006494
CAS Number: 20845-34-5
EINECS: 244-078-6
UN: 2735
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR