7-(Diethylamino)-4-methylcoumarin 99%
Supplier: Thermo Scientific Chemicals
B21938-22
B21938-36
CAAAB21938-22EA
108.39
CAD
CAAAB21938-22
CAAAB21938-36
7-(Diethylamino)-4-methylcoumarin 99%
7-(Diethylamino)-4-methylcoumarin
Purity: 99%
CAS No.: 91-44-1
Molecular Formula: C14H17NO2
Formula Weight: 231.30
Melting Point: 72-75°
Beilstein Reference: 193303
Formula:
C₁₄H₁₇NO₂ Melting Pt: 72…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006864 CAS Number: 91-44-1 EINECS: 202-068-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...