Desyl chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-2-phenylacetophenone

B21778-06 B21778-14 B21778-22
CAAAB21778-06EA 64.78 CAD
CAAAB21778-06 CAAAB21778-14 CAAAB21778-22
Desyl chloride 98%
Desyl chloride

Purity: 98% CAS No.: 447-31-4 Molecular Formula: C14H11ClO Formula Weight: 230.69 Melting Point: 64-69° Beilstein Reference: 744474 UN No.: 3261 MDL No.: MFCD00000858

Formula: C₁₄H₁₁ClO
MW: 230.69 g/mol
Melting Pt: 64…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00000858
CAS Number: 447-31-4
EINECS: 207-181-7
UN: 1759
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR