(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

B21729-03 B21729-06 B21729-14
CAAAB21729-03EA 51.51 CAD
CAAAB21729-03 CAAAB21729-06 CAAAB21729-14
(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride 97%
(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride

Purity: 97% CAS No.: 1253-47-0 Molecular Formula: C27H24ClO2P Formula Weight: 446.91 Melting Point: 230-235° MDL No.: MFCD02683069

Formula: C₂₇H₂₄ClO₂P
Melting Pt: 230…235 °C
Storage Temperature: Ambient
MDL Number: MFCD02683069
CAS Number: 1253-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR