4-Chloro-2,6-dimethoxypyrimidine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21696-06
B21696-14
CAAAB21696-03EA
107.79
CAD
CAAAB21696-03
CAAAB21696-06
CAAAB21696-14
4-Chloro-2,6-dimethoxypyrimidine 98+%
4-Chloro-2,6-dimethoxypyrimidine
Purity: 98+%
CAS No.: 6320-15-6
Molecular Formula: C6H7ClN2O2
Formula Weight: 174.59
Melting Point: 71-73°
Beilstein Reference: 137581
Formula:
C₆H₇ClN₂O₂ Melting Pt: 71…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006068 CAS Number: 6320-15-6 EINECS: 228-669-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...