2-Amino-6-bromopurine 98%

Supplier: Thermo Scientific Chemicals

B21629-03 B21629-06
CAAAB21629-03EA 66.99 CAD
CAAAB21629-03 CAAAB21629-06 CAAAB21629-14
2-Amino-6-bromopurine 98%
2-Amino-6-bromopurine

Purity: 98% CAS No.: 82499-03-4 Molecular Formula: C5H4BrN5 Formula Weight: 214.02 Melting Point: 300° MDL No.: MFCD01321309

Formula: C₅H₄BrN₅
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01321309
CAS Number: 82499-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR