Curcumin (from turmeric rhizome) 95% (total curcuminoid content)

Supplier: Thermo Scientific Chemicals
Warning

B21573-09 B21573-18 B21573-30
CAAAB21573-09EA 107.12 CAD
CAAAB21573-09 CAAAB21573-18 CAAAB21573-30
Curcumin (from turmeric rhizome) 95% (total curcuminoid content)
Curcumin

Natural Yellow 3. Inhibits lipoxygenase and cyclooxygenase


Purity: 95% CAS No.: 458-37-7 Molecular Formula: C21H20O6 Formula Weight: 368.39 Odor: Odorless Melting Point: 170-175° Beilstein Reference: 2306965 Merck Reference: 14,2673 MDL No.: MFCD00008365


Natural Yellow 3. Inhibits lipoxygenase and cyclooxygenase

Formula: C₂₁H₂₀O₆
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00008365
CAS Number: 458-37-7
EINECS: 207-280-5
Merck Index: 14,2673

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR