Diethyl phenyl orthoformate 97%

Supplier: Thermo Scientific Chemicals
Warning

B21498-09 B21498-18
CAAAAB21498-30EA 861.24 CAD
CAAAAB21498-30 CAAAB21498-09 CAAAB21498-18
Diethyl phenyl orthoformate 97%
Diethyl phenyl orthoformate

Fieser: 3,97


Purity: 97% CAS No.: 14444-77-0 Molecular Formula: C11H16O3 Formula Weight: 196.25 Flash Point: 65°(149°F) Refractive Index: 1.4810 Melting Point: -26° Boiling Point: 103-104°/10mm Beilstein Reference: 1872634 MDL No.: MFCD00009228

Formula: C₁₁H₁₆O₃
Boiling Pt: 103…104 °C/10 mm
Melting Pt: –26 °C
Density: 1.015
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009228
CAS Number: 14444-77-0
EINECS: 238-421-9
UN: 1993
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR