Diethyl terephthalate 95%

Supplier: Thermo Scientific Chemicals

B21401-09 B21401-18
CAAAB21401-09EA 58.46 CAD
CAAAB21401-09 CAAAB21401-18
Diethyl terephthalate 95%
Diethyl terephthalate

Purity: 95% CAS No.: 636-09-9 Molecular Formula: C12H14O4 Formula Weight: 222.24 Melting Point: 44-46° MDL No.: MFCD00039891

Formula: C₁₂H₁₄O₄
Melting Pt: 44…46 °C
Density: 1.100
Storage Temperature: Ambient
MDL Number: MFCD00039891
CAS Number: 636-09-9
EINECS: 211-249-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR