N-(2-Aminoethyl)morpholine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-(2-Aminoethyl)morpholine, 2-Morpholinoethanamine
B21365-0B
B21365-18
B21365-30
CAAAB21365-0BEA
213101.85
CAD
CAAAB21365-0B
CAAAB21365-18
CAAAB21365-30
N-(2-Aminoethyl)morpholine 98+%
N-(2-Aminoethyl)morpholine
Purity: 98+%
CAS No.: 2038-03-1
Molecular Formula: C6H14N2O
Formula Weight: 130.19
Flash Point: 86°(186°F)
Refractive Index: 1.4760
Odor: Amine-like
pH: 12
Melting Point: 23-25°
Boiling Point: 200-203°
Beilstein Reference: 104378
UN No.: 2923
MDL No.: MFCD00006182
Formula:
C₆H₁₄N₂O Boiling Pt: 200…203 °C Melting Pt: 23…25 °C Density: 0.999 Flash Pt: 86 °C(186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006182 CAS Number: 2038-03-1 EINECS: 218-011-6 UN: 2923 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...