Cresol red sodium salt

Supplier: Thermo Scientific Chemicals

Synonyms: Cresol Red sodium, Sodium 4-(3-(4-hydroxy-3-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-methylphenolate

B21361-14 B21361-22
CAAAB21361-14EA 127.35 CAD
CAAAB21361-14 CAAAB21361-22
Cresol red sodium salt
Cresol red sodium salt

Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red)


CAS No.: 62625-29-0 Molecular Formula: C21H17NaO5S Formula Weight: 404.42 Melting Point: ca 250° dec. MDL No.: MFCD00001618

Formula: C₂₁H₁₇NaO₅S
MW: 404.42 g/mol
Melting Pt: ∼ 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001618
CAS Number: 62625-29-0
EINECS: 263-654-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR