Dihydrocoumarin 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Chroman-2-One, 1,2-Benzodihydropyrone
B21229-0E
B21229-22
B21229-36
CAAAAB21229-0EEA
644.64
CAD
CAAAAB21229-0E
CAAAB21229-22
CAAAB21229-36
Dihydrocoumarin 99%
Dihydrocoumarin
Purity: 99%
CAS No.: 119-84-6
Molecular Formula: C9H8O2
Formula Weight: 148.16
Flash Point: 130°(266°F)
Refractive Index: 1.5560
Melting Point: 24-25°
Boiling Point: 272°
Beilstein Reference: 4584
MDL No.: MFCD00006881
Formula:
C₉H₈O₂ Boiling Pt: 272 °C Melting Pt: 24…25 °C Density: 1.169 Flash Pt: 130 °C(266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006881 CAS Number: 119-84-6 EINECS: 204-354-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...