cis,cis-1,5-Cyclooctadiene 99% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
cis,cis-Cycloocta-1,5-diene, (Z,Z)-1,5-Cyclooctadiene
Stabilization:
50-200 ppm Stabilised with IRGANOX 1076
B21196-AK
B21196-AU
CAAAB21196-AKEA
52.93
CAD
CAAAB21196-AK
CAAAB21196-AU
cis,cis-1,5-Cyclooctadiene 99% stabilised
cis,cis-1,5-Cyclooctadiene
Purity: 99%
CAS No.: 1552-12-1
Molecular Formula: C8H12
Formula Weight: 108.18
Flash Point: 32°(89°F)
Refractive Index: 1.4940
Odor: Unpleasant
Melting Point: -70°
Boiling Point: 151-152°
Beilstein Reference: 1209288
UN No.: 2520
MDL No.: MFCD00001752
Formula:
C₈H₁₂ MW: 108.18 g/mol Boiling Pt: 149 Melting Pt: –70 °C Density: 0.880 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001752 CAS Number: 1552-12-1 EINECS: 216-291-4 UN: 2520 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...