Bis(diphenylphosphino)methane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [(1,1-Diphenylphosphino)methyl](diphenyl)phosphine, DPPM

B21112-04 B21112-09 B21112-18
CAAAAB21112-04EA 45.82 CAD
CAAAAB21112-04 CAAAB21112-09 CAAAB21112-18
Bis(diphenylphosphino)methane 97%
Bis(diphenylphosphino)methane

Purity: 97% CAS No.: 2071-20-7 Molecular Formula: (C6H5)2PCH2P(C6H5)2 Formula Weight: 384.40 Melting Point: 118-119° Beilstein Reference: 758760

Formula: C₂₅H₂₂P₂
MW: 384.4 g/mol
Melting Pt: 118…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00003537
CAS Number: 2071-20-7
EINECS: 218-194-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR