1,2-Bis(dichlorophosphino)ethane 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21092-03
B21092-06
B21092-14
CAAAAB21092-03EA
119.76
CAD
CAAAAB21092-03
CAAAAB21092-06
CAAAAB21092-14
1,2-Bis(dichlorophosphino)ethane 96%
1,2-Bis(dichlorophosphino)ethane
Purity: 96%
CAS No.: 28240-69-9
Molecular Formula: Cl2PCH2CH2PCl2
Formula Weight: 231.81
Flash Point: None
Refractive Index: 1.5880
Boiling Point: 70°/1mm
UN No.: 3265
MDL No.: MFCD00009609
Formula:
C₂H₄Cl₄P₂ MW: 231.813 g/mol Boiling Pt: 70 °C (1 mmHg) Density: 1.54 Storage Temperature: Ambient |
MDL Number:
MFCD00009609 CAS Number: 28240-69-9 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...