Cyclohexyl methyl ketone 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Cyclohexylethanone

B21000-06 B21000-14 B21000-22
CAAAB21000-06EA 74.1 CAD
CAAAB21000-06 CAAAB21000-14 CAAAB21000-22
Cyclohexyl methyl ketone 95%
Cyclohexyl methyl ketone

Purity: 95% CAS No.: 823-76-7 Molecular Formula: C8H14O Formula Weight: 126.20 Flash Point: 66°(151°F) Physical Form: Liquid Density: 0.917 Refractive Index: 1.4520 Melting Point: -34° Boiling Point: 181-183° Beilstein Reference: 507229 MDL No.: MFCD00040418

Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 181…183 °C
Melting Pt: –34 °C
Density: 0.917
Flash Pt: 66 °C(151 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040418
CAS Number: 823-76-7
EINECS: 212-517-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR