Chloro(di-tert-butyl)phosphine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Di-tert-butylchlorophosphine, Di-t-butylchlorophosphine

B20998-06 B20998-14
CAAAAB20998-06EA 186.44 CAD
CAAAAB20998-06 CAAAAB20998-14
Chloro(di-tert-butyl)phosphine 96%
Chloro(di-tert-butyl)phosphine

Purity: 96% CAS No.: 13716-10-4 Molecular Formula: C8H18ClP Formula Weight: 180.66 Flash Point: 62°(144°F) Refractive Index: 1.4840 Odor: Acrid Boiling Point: 47-49°/3mm UN No.: 3265 MDL No.: MFCD00008815

Formula: C₈H₁₈ClP
MW: 180.66 g/mol
Boiling Pt: 47…49 °C (3 mmHg)
Density: 0.951
Flash Pt: 62 °C (144 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008815
CAS Number: 13716-10-4
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR