(±)-2-Methyltetrahydrofuran-3-one 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Methyltetrahydrofuran-3-one
B20890-06
B20890-14
CAAAB20890-06EA
74.58
CAD
CAAAB20890-06
CAAAB20890-14
(±)-2-Methyltetrahydrofuran-3-one 98+%
(±)-2-Methyltetrahydrofuran-3-one
Purity: 98+%
CAS No.: 3188-00-9
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 39°(102°F)
Refractive Index: 1.4290
Odor: Characteristic
Boiling Point: 138-139°
Beilstein Reference: 1341334
UN No.: 1224
MDL No.: MFCD00010423
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 138…139 °C Density: 1.034 Flash Pt: 39 °C (102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010423 CAS Number: 3188-00-9 EINECS: 221-685-4 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...