Methyl-3-ethoxythiophene-2-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20860-03
B20860-06
CAAAB20860-03EA
102.38
CAD
CAAAB20860-03
CAAAB20860-06
Methyl-3-ethoxythiophene-2-carboxylate 97%
Methyl 3-ethoxythiophene-2-carboxylate
Purity: 97%
CAS No.: 139926-22-0
Molecular Formula: C8H10O3S
Formula Weight: 186.23
Melting Point: 72-74°
MDL No.: MFCD00102116
Formula:
C₈H₁₀O₃S Melting Pt: 72…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00102116 CAS Number: 139926-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...