Rubidium tetrafluoroborate 98%

Supplier: Thermo Scientific Chemicals
Warning

B20817-06 B20817-14
CAAAAB20817-06EA 96.7 CAD
CAAAAB20817-06 CAAAAB20817-14
Rubidium tetrafluoroborate 98%
Rubidium tetrafluoroborate

Purity: 98% CAS No.: 18909-68-7 Molecular Formula: BF4Rb Formula Weight: 172.27 MDL No.: MFCD00083071

Formula: BF₄Rb
Storage Temperature: Ambient
MDL Number: MFCD00083071
CAS Number: 18909-68-7
EINECS: 242-660-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR