Methyl-6-methoxynicotinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 6-methoxynicotinate, 6-Methoxynicotinic acid methylester, Methyl 2-methoxy-5-pyridinecarboxylate

B20808-09 B20808-18
CAAAB20808-09EA 99.38 CAD
CAAAB20808-09 CAAAB20808-18 CAAAB20808-30
Methyl-6-methoxynicotinate 98%
Methyl 6-methoxynicotinate

Purity: 98% CAS No.: 26218-80-4 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: ca 48-53° MDL No.: MFCD00674130

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: ca 48…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00674130
CAS Number: 26218-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR