Ethyl-5-bromonicotinate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20788-14
B20788-06
CAAAAB20788-14EA
1196.06
CAD
CAAAAB20788-14
CAAAB20788-03
CAAAB20788-06
Ethyl-5-bromonicotinate 98%
Ethyl 5-bromonicotinate
Purity: 98%
CAS No.: 20986-40-7
Molecular Formula: C8H8BrNO2
Formula Weight: 230.07
Melting Point: 39-42°
Boiling Point: 86-92°/0.5mm
MDL No.: MFCD00040366
Formula:
C₈H₈BrNO₂ Boiling Pt: 86…92 °C/0.5 mm Melting Pt: 39…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040366 CAS Number: 20986-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...