Methyl-5-bromo-2-furoate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2-furoic acid methyll ester, 5-Bromo-2-furancarboxylic acid methyll ester, Methyl 5-Bromo-2-furancarboxylate, Methyl 5-Bromo-2-furoate, Methyl 5-bromofuran-2-carboxylate

B20524-03 B20524-06
CAAAB20524-03EA 44.71 CAD
CAAAB20524-03 CAAAB20524-06
Methyl-5-bromo-2-furoate 97%
Methyl 5-bromo-2-furoate

Purity: 97% CAS No.: 2527-99-3 Molecular Formula: C6H5BrO3 Formula Weight: 205.01 Melting Point: 62-65° MDL No.: MFCD00092313

Formula: C₆H₅BrO₃
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00092313
CAS Number: 2527-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR