Cyclohexyl ethyl ketone 99%

Supplier: Thermo Scientific Chemicals

B20409-06 B20409-14
CAAAB20409-06EA 366.56 CAD
CAAAB20409-06 CAAAB20409-14
Cyclohexyl ethyl ketone 99%
Cyclohexyl ethyl ketone

Purity: 99% CAS No.: 1123-86-0 Molecular Formula: C9H16O Formula Weight: 140.23 MDL No.: MFCD00060814

Formula: C₉H₁₆O
Density: 0.91
Storage Temperature: Ambient
MDL Number: MFCD00060814
CAS Number: 1123-86-0
EINECS: 214-380-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR