Ethyl-2-chloronicotinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-Chloro-3-pyridinecarboxylate, Ethyl 2-chloronicotinate

B20359-09 B20359-18
CAAAB20359-09EA 130.19 CAD
CAAAB20359-09 CAAAB20359-18 CAAAB20359-30
Ethyl-2-chloronicotinate 98%
Ethyl 2-chloronicotinate

Purity: 98% CAS No.: 1452-94-4 Molecular Formula: C8H8ClNO2 Formula Weight: 185.61 MDL No.: MFCD00674129

Formula: C₈H₈ClNO₂
MW: 185.61
Storage Temperature: Ambient
MDL Number: MFCD00674129
CAS Number: 1452-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR