4-Hydrazinobenzoic acid hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20326-09
B20326-18
CAAAB20326-09EA
64.46
CAD
CAAAB20326-09
CAAAB20326-18
CAAAB20326-30
4-Hydrazinobenzoic acid hydrochloride 98%
4-Hydrazinobenzoic acid hydrochloride
Purity: 98%
CAS No.: 24589-77-3
Molecular Formula: C7H8N2O2.HCl
Formula Weight: 188.61
Melting Point: 252-254° dec.
MDL No.: MFCD00039073
Formula:
C₇H₉ClN₂O₂ Melting Pt: 252…254 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00039073 CAS Number: 24589-77-3 EINECS: 246-330-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...