3-Ethoxy-2-thenoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Ethoxythiophene-2-carboxylic acid

B20296-03 B20296-06
CAAAB20296-03EA 150.1 CAD
CAAAB20296-03 CAAAB20296-06 CAAAB20296-14
3-Ethoxy-2-thenoic acid 97%
3-Ethoxy-2-thenoic acid

Purity: 97% CAS No.: 139926-23-1 Molecular Formula: C7H8O3S Formula Weight: 172.20 Melting Point: 121-123° MDL No.: MFCD00205205

Formula: C₇H₈O₃S
MW: 172.2
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00205205
CAS Number: 139926-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR