8-Hydroxyquinoline-5-sulfonic acid hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

B20022-36 B20022-18 B20022-30
CAAAAB20022-36EA 301.78 CAD
CAAAAB20022-36 CAAAB20022-18 CAAAB20022-30
8-Hydroxyquinoline-5-sulfonic acid hydrate 98%
8-Hydroxyquinoline-5-sulphonic acid hydrate

Purity: 98% CAS No.: 207386-92-3 Molecular Formula: C9H7NO4S.xH2O Formula Weight: 225.22(anhy) Melting Point: 311-313° Beilstein Reference: 195247 Merck Reference: 14,4844 MDL No.: MFCD00149482

Formula: C₉H₇NO₄S·H₂O
MW: 225.22 g/mol (anhy)
Melting Pt: 311…313 °C
Storage Temperature: Ambient
MDL Number: MFCD00149482
CAS Number: 207386-92-3
EINECS: 201-570-5
Merck Index: 14,4844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR