Sodium-tert-butanolate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate
A19942-0E
A19942-22
A19942-36
CAAAAA19942-0EEA
1294.02
CAD
CAAAAA19942-0E
CAAAAA19942-22
CAAAAA19942-36
Sodium-tert-butanolate 97%
Sodium tert-butanolate
Purity: 97%
CAS No.: 865-48-5
Molecular Formula: C4H9NaO
Formula Weight: 96.11
Odor: Odorless
Beilstein Reference: 3654215
UN No.: 3206
MDL No.: MFCD00040575
Formula:
C₄H₉NaO MW: 96.1 g/mol Density: 0.785 Storage Temperature: Ambient |
MDL Number:
MFCD00040575 CAS Number: 865-48-5 EINECS: 212-741-9 UN: 3206 ADR: "4.2",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...