2-Bromo-6-methoxynaphthalene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19450-09
A19450-18
CAAAA19450-09EA
85.95
CAD
CAAAA19450-09
CAAAA19450-18
2-Bromo-6-methoxynaphthalene 98%
2-Bromo-6-methoxynaphthalene
Purity: 98%
CAS No.: 5111-65-9
Molecular Formula: C11H9BrO
Formula Weight: 237.10
Melting Point: 105-110°
Boiling Point: 160-164°/3mm
MDL No.: MFCD00004062
Formula:
C₁₁H₉BrO Boiling Pt: 160…164 °C/3 mm Melting Pt: 105…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004062 CAS Number: 5111-65-9 EINECS: 225-837-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...