2-(Bromomethyl)-1,3-dioxolane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19312-14
A19312-22
CAAAA19312-14EA
110.44
CAD
CAAAA19312-14
CAAAA19312-22
2-(Bromomethyl)-1,3-dioxolane 97%
2-(Bromomethyl)-1,3-dioxolane
Purity: 97%
CAS No.: 4360-63-8
Molecular Formula: C4H7BrO2
Formula Weight: 167.01
Flash Point: 62°(145°F)
Refractive Index: 1.4820
Boiling Point: 175°
Beilstein Reference: 103309
MDL No.: MFCD00003214
Formula:
C₄H₇BrO₂ Boiling Pt: 175 °C Density: 1.628 Flash Pt: 62 °C(145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003214 CAS Number: 4360-63-8 EINECS: 224-443-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...