3-Methyl-2-thenoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methylthiophene-2-carboxylic acid

A19194-22 A19194-06 A19194-14
CAAAAA19194-22EA 717.32 CAD
CAAAAA19194-22 CAAAA19194-06 CAAAA19194-14
3-Methyl-2-thenoic acid 98%
3-Methyl-2-thenoic acid

Purity: 98% CAS No.: 23806-24-8 Molecular Formula: C6H6O2S Formula Weight: 142.18 Melting Point: 144-149° Beilstein Reference: 116184

Formula: C₆H₆O₂S
MW: 142.18 g/mol
Melting Pt: 144…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00005438
CAS Number: 23806-24-8
EINECS: 245-894-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR